Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ CDK12, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABE1861MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABE1861MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1861MI Supplier MilliporeSigma™ Supplier No. ABE1861
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cyclin-dependent kinase 12 (EC 2.7.11.22; EC 2.7.11.23; UniProt Q9NYV4; also known as CDC2-related protein kinase 7, Cdc2-related kinase, arginine/serine-rich, Cell division cycle 2-related protein kinase 7, Cell division protein kinase 12, CrkRS, hCDK12) is encoded by the CDK12 (also known as CRK7, CRKRS, KIAA0904) gene (Gene ID 51755) in human. CDK12 belongs to the CMGC Ser/Thr protein kinase subfamily of cyclin-dependent kinases (CDKs). Originally identified and characterized as an RNA Polymerase II C-terminal domain (CTD) kinase, CDK12 is involved in the 3' end processing of the MYC and c-FOS mRNA transcripts. In addtion, CDK12/CyclinK complex is implicated in the maintenance of genomic stability through the regulation of DNA damage response genes and as a determinant of PARP1/2 inhibitor sensitivity in cancer treatment. Human CDK12 is uncharacteristically large with a molecular weight of ∽160kDa (most CDKs are about 50kDa)and contains large N- and C-terminal extensions flanking the kinase domain (a.a. 727-1020). These extensions contain multiple stretches of low complexity sequence and RS domains characteristic of splicing factors and regulators believed to mediate protein-protein interactions. As expected of RS domain-containing proteins, CDK12 is found in nuclear speckles representing storage sites for splicing factors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDK12
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene CDK12, CRK7, CRKRS, KIAA0904
Gene Accession No. Q9NYV4
Host Species Rabbit
Immunogen Linear peptide corresponding to an N-terminal sequence of human CDK12.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_057591
Test Specificity Immunogen sequence is present in all spliced isoforms of human, mouse, and rat CDK12 reported by UniProt (Q9NYV4, Q14AX6, and Q3MJK5).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.