Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDK13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563692
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563692 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563692 Supplier Invitrogen™ Supplier No. PA563692
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PLGGIQPSSQ TIQPKVETDA AQAAVQSAFA VLLTQLIKAQ QSKQKDVLLE ERENGSGHEA SLQLRPPPEP STPVSGQDDL IQHQDMRILE LTPEPDRPRI LPPDQRPPE Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 89%.

CDK13/cyclin K serine/threonine kinase is dimeric protein complex. CDK13 is a member of the cyclin-dependent serine/threonine protein kinase family. Members of this family are well known for their essential roles as master switches in cell cycle control. The exact function of this protein has not yet been determined, but it may play a role in mRNA processing and may be involved in regulation of hematopoiesis. Cyclin K is a member of the transcription cyclin family. These cyclins may regulate transcription through their association with and activation of cyclin-dependent kinases (CDK) that phosphorylate the C-terminal domain of the large subunit of RNA polymerase II.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDK13
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Cdk13
Gene Accession No. Q14004
Gene Alias 2310015O17Rik; CDC2L; Cdc2l5; CDC2-related protein kinase 5; CDK13; cell division cycle 2-like 5 (cholinesterase-related cell division controller); cell division cycle 2-like protein kinase 5; Cell division protein kinase 13; CHED; Cholinesterase-related cell division controller; cyclin dependent kinase 13; cyclin-dependent kinase 13; hCDK13; Kiaa1791
Gene Symbols Cdk13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CDK13. Recombinant protein control fragment (Product #RP-95227).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8621
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.