Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CDK18/cyclin Y, GST Tag Recombinant Protein

Catalog No. A33352
Encompass_Preferred
Click to view available options
Quantity:
1 mg
10 μg
100 μg

Recombinant Protein

CDK18/cyclin Y serine/threonine kinase or cyclin-dependent kinase 18 belongs to the cdc2/cdkx subfamily of kinases. CDK18/cyclin Y is thought to play a role in terminally differentiated neurons and it contains an N-terminal extension relative to CDC2. CDK18/cyclin Y is expressed in a variety of human cell lines and tissues. Elevation in the protein levels of CDK18/cyclin Y in the temporal cortex of Alzheimer's disease relative to control brains has been observed. It is proposed that CDK18/cyclin Y is a paired helical filaments-associated kinase that modulates tau phosphorylation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q07002
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 5129
Molecular Weight (g/mol) 80.5-65.5 kDa
Name Human CDK18/cyclin Y, GST Tag
Quantity 1 mg
Gene Alias PCTK3
Common Name CDK18
KinaseFamily CDKs and CHKs
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence CDK18: Full Length, Cyclin Y: 2-341
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Form Recombinant, Active, Catalytic
Form Liquid
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.