Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDK2 Antibody (C15.2), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002686H
Description
CDK2 Monoclonal specifically detects CDK2 in Xenopus samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
CDK2 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
CDK2 | |
Xenopus Cdk2 | |
0.1 mL | |
Breast Cancer, Cancer, Cell Cycle and Replication, Ovarian Carcinoma Cell Markers | |
1017 | |
Store at 4C in the dark. | |
IgG2a |
Western Blot, ELISA, Immunohistochemistry | |
C15.2 | |
Western Blot, ELISA, Immunohistochemistry | |
cdc2-related protein kinase, cell devision kinase 2, Cell division protein kinase 2, cyclin-dependent kinase 2, EC 2.7.11, EC 2.7.11.22, p33 protein kinase, p33(CDK2) | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Xenopus species | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction