Learn More
Promega CDK2/CyclinA2 Kinase Enzyme System, 1mg
Catalog No. PRV2972 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV2972 1 mg
1 options
Catalog No. PRV2972 Supplier Promega Supplier No. V2972
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human CDK2 and CyclinA2 were co-expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag on both proteins. CDK2 is a member of the Cyclin-Dependent Kinase family that is ubiquitously expressed.
  • Easily Screen and Profile CDK2/CyclinA2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C