Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDK2 Monoclonal Antibody (10-D7)
GREENER_CHOICE

Catalog No. PIMA532903
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532903 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532903 Supplier Invitrogen™ Supplier No. MA532903
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CDK2 Monoclonal Antibody for ICC/IF, Western Blot

CDK2/cyclinO is a member of the cyclin dependent protein kinase family complexed to CyclinO. CyclinO was used as a specific protein isoform name for the UNG gene which is localized exclusively in the nucleus. In addition, CyclinO was reported to shows significant uracil-DNA glycosylase activity. CDK2/cyclinO protein levels increase during G1 phase and the protein is turned over during the course of cell cycle. The levels of cyclin A-cdk2 is maximal at the G1/S transition and both cdk2 and cyclin A are found associated with DNA in the initiation complex during replication. Rb protein acts as substrate for cdk2-cyclin E and/or cdk2-cyclin A in vivo. Cdk2 is activated and specifically localized to the nucleus during late G1-, S-, and G2-phase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDK2
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 10-D7
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% Glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Gene CDK2
Gene Accession No. P24941, P97377, Q63699
Gene Alias A630093N05Rik; CDC28; CDC2A; cdc2-related protein kinase; CDK1; CDK2; CDKN2; Cell division protein kinase 2; cyclin dependent kinase 2; cyclin dependent kinase 2-alpha; cyclin-dependent kinase 2; EC 2.7.11.22; EC 2.7.11.23; kinase Cdc2; MPF; p33 protein kinase; p33(CDK2); p33CDK2; p34 protein kinase
Gene Symbols CDK2
Host Species Mouse
Immunogen Synthetic peptide within Human CDK2 aa 249-298.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1017, 12566, 362817
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.