Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDK2 Monoclonal Antibody (5B12D1), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA549149

Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Human Cdk2 shares 98.6% amino acid (aa) sequence identity with rat Cdk2.
CDK2/cyclinO is a member of the cyclin dependent protein kinase family complexed to CyclinO. CyclinO was used as a specific protein isoform name for the UNG gene which is localized exclusively in the nucleus. In addition, CyclinO was reported to shows significant uracil-DNA glycosylase activity. CDK2/cyclinO protein levels increase during G1 phase and the protein is turned over during the course of cell cycle. The levels of cyclin A-cdk2 is maximal at the G1/S transition and both cdk2 and cyclin A are found associated with DNA in the initiation complex during replication. Rb protein acts as substrate for cdk2-cyclin E and/or cdk2-cyclin A in vivo. Cdk2 is activated and specifically localized to the nucleus during late G1-, S-, and G2-phase.
Specifications
CDK2 | |
Monoclonal | |
500 μg/mL | |
PBS with 4MG trehalose and no preservative | |
P24941, P97377, Q63699 | |
CDK2 | |
E.coli-derived human Cdk2 recombinant protein (Position: E81-L298). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
5B12D1 | |
Unconjugated | |
CDK2 | |
A630093N05Rik; CDC28; CDC2A; cdc2-related protein kinase; CDK1; CDK2; CDKN2; Cell division protein kinase 2; cyclin dependent kinase 2; cyclin dependent kinase 2-alpha; cyclin-dependent kinase 2; EC 2.7.11.22; EC 2.7.11.23; kinase Cdc2; MPF; p33 protein kinase; p33(CDK2); p33CDK2; p34 protein kinase | |
Mouse | |
Antigen affinity chromatography | |
RUO | |
1017, 12566, 362817 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction