Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDK2 Monoclonal Antibody (OTI1A8), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502943

Description
p33cdk2 associates with cyclin A in human cells. Kinase activity associated with cyclin A-cdc2 is found only in G2-phase. Cdk2 also complexes with cyclins E, D1, and D3. Cyclin E-cdk2 kinase is active in the G1- and S-phases of the cell cycle and is important (as does cyclin A-cdk2) for the progression from G1-to S-phase. The levels of cyclin A-cdk2 is maximal at the G1/S transition and both cdk2 and cyclin A are found associated with DNA in the initiation complex during replication. Rb protein acts as substrate for cdk2-cyclin E and/or cdk2-cyclin A in vivo. Cdk2 is activated and specifically localized to the nucleus during late G1-, S-, and G2-phase.Specifications
CDK2 | |
Monoclonal | |
0.86 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P24941 | |
CDK2 | |
Full length human recombit protein of human CDK2 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1A8 | |
Unconjugated | |
CDK2 | |
CDKN2, p33(CDK2) | |
Mouse | |
Affinity Chromatography | |
RUO | |
1017 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction