Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDK2, p Tyr15 Antibody (2C4), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | CDK2 |
---|---|
Clone | 2C4 |
Dilution | Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:200-1:1000 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation |
Classification | Monoclonal |
Description
CDK2 Monoclonal antibody specifically detects CDK2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, ImmunoprecipitationSpecifications
CDK2 | |
Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:200-1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
cdc2-related protein kinase, cell devision kinase 2, Cell division protein kinase 2, cyclin-dependent kinase 2, EC 2.7.11, EC 2.7.11.22, p33 protein kinase, p33(CDK2) | |
A synthesized peptide derived from Human CDK2 [p Tyr15] [UniProt P24941] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
2C4 | |
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Breast Cancer, Cancer, Cell Cycle and Replication, Ovarian Carcinoma Cell Markers, Phospho Specific | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
1017 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title