Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CDK20 Monoclonal Antibody (885CT27.1.1)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA537622
Description
CDK20 Monoclonal Antibody for Western Blot
The protein encoded by this gene contains a kinase domain most closely related to the cyclin-dependent protein kinases. The encoded kinase may activate cyclin-dependent kinase 2 and is involved in cell growth. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications
CDK20 | |
Monoclonal | |
0.4 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q8IZL9 | |
CDK20 | |
purified His-tagged CCRK protein. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
885CT27.1.1 | |
Unconjugated | |
CDK20 | |
4932702G04Rik; AU019450; AW558027; CAK-kinase p42; Ccrk; CDCH; CDK20; CDK-activating kinase p42; CDK-related protein kinase PNQLARE; cell cycle related kinase; cell cycle-related kinase; Cell division protein kinase 20; cyclin dependent kinase 20; cyclin-dependent kinase 20; Cyclin-dependent protein kinase H; cyclin-kinase-activating kinase p42; p42; PNQALRE; PNQLARE | |
Mouse | |
Protein G | |
RUO | |
23552 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction