Learn More
CDK20 Mouse anti-Human, Clone: OTI1B3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809548

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene contains a kinase domain most closely related to the cyclin-dependent protein kinases. The encoded kinase may activate cyclin-dependent kinase 2 and is involved in cell growth. Alternatively spliced transcript variants encoding distinct isoforms have been reported.Specifications
CDK20 | |
Monoclonal | |
Unconjugated | |
CDK20 | |
CCRK, CDCH, P42, PNQALRE | |
Mouse | |
Affinity Chromatography | |
RUO | |
23552 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1B3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8IZL9 | |
CDK20 | |
Full length human recombit protein of human CDK20 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.