Learn More
Invitrogen™ CDK20 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588163
Description
Immunogen sequence: MDQYCILGRI GEGAHGIVFK AKHVETGEIV ALKKVALRRL EDGFPNQALR EIKALQEMED NQYVVQLKAV FPHGGGFVLA FEFMLSDLAE VVRHAQRPLA QAQVKSYLQM LLKGVAFCHA NNIVHRDLKP ANLLISASGQ LKIADFGLAR VFSPDGSRLY THQVATRSVG CIMGELLNGS PLFPGKNDIE QLCYVLRILG TPNPQVWPEL TELPDYNKIS FKEQVPMPLE EVLPDVSPQA LDLLGQFLLY PPHQRIAASK LPCLPIHLSC RFLSV; Positive Samples: HeLa, U-87MG, HepG2, Mouse heart, Mouse eye; Cellular Location: Cell projection, Cytoplasm, Nucleus, cilium.
The protein encoded by this gene contains a kinase domain most closely related to the cyclin-dependent protein kinases. The encoded kinase may activate cyclin-dependent kinase 2 and is involved in cell growth. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications
CDK20 | |
Polyclonal | |
Unconjugated | |
CDK20 | |
4932702G04Rik; AU019450; AW558027; CAK-kinase p42; Ccrk; CDCH; CDK20; CDK-activating kinase p42; CDK-related protein kinase PNQLARE; cell cycle related kinase; cell cycle-related kinase; Cell division protein kinase 20; cyclin dependent kinase 20; cyclin-dependent kinase 20; Cyclin-dependent protein kinase H; cyclin-kinase-activating kinase p42; p42; PNQALRE; PNQLARE | |
Rabbit | |
Affinity Chromatography | |
RUO | |
105278, 23552 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.51 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q8IZL9, Q9JHU3 | |
CDK20 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-275 of human CDK20 (NP_0011641101). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.