Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDK2AP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP25692425UL
Description
CDK2AP1 Polyclonal specifically detects CDK2AP1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
CDK2AP1 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
CDK2-associated protein 1cyclin-dependent kinase 2-associated protein 1, CDKAP1, cyclin-dependent kinase 2 associated protein 1, Deleted in oral cancer 1, Deleted in oral cancer-1, doc-1, DOC1Putative oral cancer suppressor, DORC1, p12DOC-1, ST19 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
IgG |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
CDK2AP1 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MSYKPNLAAHMPAAALNAAGSVHSPSTSMA | |
25 μL | |
Breast Cancer, Cancer | |
8099 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction