Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDK2AP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144782
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144782 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144782 Supplier Invitrogen™ Supplier No. PA5144782
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SPSGSVPGAG APFRPLFNDF GPPSMGYVQA MKPPGAQGSQ STYTDLLSVI EEMG Antigen sequence identity: Mouse 96%; Rat 96%.

This gene encodes a protein that interacts with cyclin-dependent kinase 2 associated protein 1. Pseudogenes associated with this gene are located on chromosomes 7 and 9. Alternatively spliced transcript variants have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDK2AP2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDK2AP2
Gene Accession No. O75956
Gene Alias 5830466O21Rik; Cdk2ap2; CDK2-associated protein 2; cyclin dependent kinase 2 associated protein 2; cyclin-dependent kinase 2 associated protein 2; cyclin-dependent kinase 2-associated protein 2; D19Ertd144e; DOC-1 related protein; Doc1r; DOC-1R; DOC-1-related protein; p14; tumor suppressor deleted in oral cancer related 1
Gene Symbols CDK2AP2
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 32-85. Recombinant protein control fragment (Product #RP-97455).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10263
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.