Learn More
Promega CDK3/CyclinE1 Kinase Enzyme System, 10microg
Catalog No. PRV4490 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4490 10 μg
1 options
Catalog No. PRV4490 Supplier Promega Supplier No. V4490
Recombinant full-length human CDK3 and Cyclin E1 were co-expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag on both proteins. CDK3/CyclinE1 is a member of the cyclin-dependent protein kinase family.
  • Easily Screen and Profile CDK3/CyclinE1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C