Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDK4 Antibody (6D10), Alexa Fluor™ 488, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP242642AF488

Catalog No. NBP242642AG


Only null left
Add to Cart

Description

Description

CDK4 Monoclonal specifically detects CDK4 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

CDK4
Monoclonal
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
CDK4
Recombinant human CDK4 (1-303aa) purified from E. coli
100 μL
Cancer, Cell Cycle and Replication, Core ESC Like Genes, mTOR Pathway, Protein Kinase, Stem Cell Markers
1019
Store at 4C in the dark.
IgG2a κ
Western Blot, ELISA
6D10
Western Blot, ELISA
Cell division protein kinase 4, CMM3, cyclin-dependent kinase 4, EC 2.7.11, EC 2.7.11.22, melanoma cutaneous malignant, 3, MGC14458, PSK-J3cell division kinase 4
Mouse
Protein A purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.