Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDK4 Antibody (8F2), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | CDK4 |
---|---|
Clone | 8F2 |
Dilution | Western Blot 1:500-1:5000, ELISA, Immunoprecipitation 1:200-1:1000 |
Applications | Western Blot, ELISA, Immunoprecipitation |
Classification | Monoclonal |
Description
CDK4 Monoclonal antibody specifically detects CDK4 in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
CDK4 | |
Western Blot 1:500-1:5000, ELISA, Immunoprecipitation 1:200-1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Cell division protein kinase 4, CMM3, cyclin-dependent kinase 4, EC 2.7.11, EC 2.7.11.22, melanoma cutaneous malignant, 3, MGC14458, PSK-J3cell division kinase 4 | |
A synthesized peptide derived from Human CDK4 [UniProt P11802] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
8F2 | |
Western Blot, ELISA, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Cancer, Cell Cycle and Replication, Core ESC Like Genes, mTOR Pathway, Protein Kinase, Stem Cell Markers | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
1019 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title