Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdk4 Mouse anti-Human, Clone: 2F6, Bioss
SDP

Catalog No. 501989533
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9533 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9533 Supplier Bioss Supplier No. BSM52028M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CDK4 is a member of the Ser/Thr protein kinase family. This protein is highly similar to the gene products of S. cerevisiae cdc28 and S. pombe cdc2. It is a catalytic subunit of the protein kinase complex that is important for cell cycle G1 phase progression. The activity of this kinase is restricted to the G1-S phase, which is controlled by the regulatory subunits D-type cyclins and CDK inhibitor p16(INK4a). This kinase was shown to be responsible for the phosphorylation of retinoblastoma gene product (Rb). Mutations in this gene as well as in its related proteins including D-type cyclins, p16(INK4a) and Rb were all found to be associated with tumorigenesis of a variety of cancers. Multiple polyadenylation sites of the gene have been reported.

Specifications

Antigen Cdk4
Applications Western Blot
Classification Monoclonal
Clone 2F6
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 40% glycerol and 0.05% sodium azide; pH
Gene Cdk4
Gene Accession No. P11802
Gene Alias BOS_5594; CDK4; Cell division protein kinase 4; CMM3; Crk3; cyclin dependent kinase 4; Cyclin-dependent kinase 4; MGC14458; p34/cdk4; PSK-J3; serine/threonine kinase
Gene Symbols Cdk4
Host Species Mouse
Immunogen Recombinant human Cdk4 protein, around C-terminal 101aa.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1019
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2c
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.