Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDK4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 110261AP150UL
Description
Cdk4 is involved in the control of the cell cycle.Specifications
| CDK4 | |
| Polyclonal | |
| Unconjugated | |
| CDK4 | |
| CDK4, Cell division protein kinase 4, CMM3, cyclin dependent kinase 4, PSK J3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1019, 12567, 94201 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P11802, P30285, P35426 | |
| CDK4 | |
| CDK4 Fusion Protein Ag1505 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction