Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cdk5 Antibody (DC17+DC34), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP323218
Description
Cdk5 Monoclonal antibody specifically detects Cdk5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, ImmunoprecipitationSpecifications
Cdk5 | |
Monoclonal | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunofluorescence, Immunoprecipitation | |
DC17+DC34 | |
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:100 | |
Cell division protein kinase 5, cyclin-dependent kinase 5, EC 2.7.11, EC 2.7.11.22, protein kinase CDK5 splicing, PSSALRE, Serine/threonine-protein kinase PSSALRE, Tau protein kinase II catalytic subunit, TPKII catalytic subunit | |
Clones DC17+DC34 were generated from a recombinant full-length human Cdk5 sequence. [UniProt# Q00535] | |
0.1 mL | |
Cell Cycle and Replication, Innate Immunity, Phospho Specific | |
1020 | |
Store at -20°C. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction