Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cdk5 Antibody (OTI2H6), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270396PE
Description
Cdk5 Monoclonal specifically detects Cdk5 in Human, Mouse, Rat, Canine samples. It is validated for Flow Cytometry.Specifications
Cdk5 | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
CDK5 | |
Full length recombinant protein of human Cdk5 (NP_004926) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine | |
IgG1 |
Flow Cytometry | |
OTI2H6 | |
Flow Cytometry | |
Cell division protein kinase 5, cyclin-dependent kinase 5, EC 2.7.11, EC 2.7.11.22, protein kinase CDK5 splicing, PSSALRE, Serine/threonine-protein kinase PSSALRE, Tau protein kinase II catalytic subunit, TPKII catalytic subunit | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication, Innate Immunity, Phospho Specific | |
1020.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction