Learn More
Invitrogen™ CDK5 Monoclonal Antibody (DC17+DC34)

Description
The antibody detects a 31 kDa* band corresponding to Cdk5 on SDS-PAGE immunoblots of human SYF and HeLa cells, and in mouse brain tissue. The antibody does not detect other Cdk family members, such as Cdk1 and Cdk2.
Specifications
Specifications
| Antigen | CDK5 |
| Applications | Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | DC17+DC34 |
| Concentration | ~0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide |
| Gene | CDK5 |
| Gene Accession No. | P49615, Q00535, Q03114 |
| Gene Alias | AW048668; Cdk5; CDK5 activator 1; CDK5-SV; CDKN5; Cell division protein kinase 5; CR6 protein kinase; Crk6; cyclin dependent kinase 5; cyclin-dependent kinase 5; cyclin-dependent-like kinase 5; LIS7; protein kinase CDK5 splicing; PSSALRE; Serine/threonine-protein kinase PSSALRE; tau protein kinase II catalytic subunit; TPKII catalytic subunit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.