Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cdk5, p Tyr15 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP312917
This item is not returnable.
View return policy
Description
Cdk5 Polyclonal antibody specifically detects Cdk5 in Human samples. It is validated for Western BlotSpecifications
Cdk5 | |
Polyclonal | |
Western Blot 1:1000-1:10000 | |
Cell division protein kinase 5, cyclin-dependent kinase 5, EC 2.7.11, EC 2.7.11.22, protein kinase CDK5 splicing, PSSALRE, Serine/threonine-protein kinase PSSALRE, Tau protein kinase II catalytic subunit, TPKII catalytic subunit | |
Carrier-protein conjugated synthetic peptide corresponding to residues around human Cdk5 (phospho Tyr15). The exact sequence is proprietary. | |
100 μg | |
Cell Cycle and Replication, Innate Immunity, Phospho Specific | |
1020 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
PBS, 1% BSA, 20% Glycerol (pH7) | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction