Learn More
Promega CDK5/p35 Kinase Enzyme System, 10microg
Catalog No. PRV3271 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV3271 10 μg
1 options
Catalog No. PRV3271 Supplier Promega Supplier No. V3271
Recombinant full-length human CDK5 and human p35 were co-expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CDK5 is a member of the Cyclin-Dependent Kinase family that is most abundant in the mammalian brain.
  • Easily Screen and Profile CDK5/p35 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C