Learn More
Promega CDK5/p35 Kinase Enzyme System, 1mg
Catalog No. PRV3272 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV3272 1 mg
1 options
Catalog No. PRV3272 Supplier Promega Supplier No. V3272
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human CDK5 and human p35 were co-expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CDK5 is a member of the Cyclin-Dependent Kinase family that is most abundant in the mammalian brain.
  • Easily Screen and Profile CDK5/p35 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C