Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDK5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120609
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120609 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120609 Supplier Invitrogen™ Supplier No. PA5120609
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: 293T. The target is usually found in the following locations: Cell junction, Cell membrane, Cell projection, Cytoplasm, Nucleus, Perikaryon, Peripheral membrane protein, growth cone, lamellipodium, postsynaptic cell membrane, postsynaptic density, synapse. Immunogen sequence: PDVLFGAKLY STSIDMWSAG CIFAELANAG RPLFPGNDVD DQLKRIFRLL GTPTEEQWPS MTKLPDYKPY PMYPATTSLV NVVPKLNATG RDLLQNLLKC NPVQRISAEE ALQHPYFSDF.

CDK5 is a member of the Cyclin-Dependent Kinase family that is most abundant in the mammalian brain. Active form of CDK5, which has also been called neuronal cdc2-like kinase, is a heterodimer of CDK5 and a 25 kDa protein which is derived proteolytically from a 35 kDa brain and neuron-specific protein and is essential for the kinase activity of CDK5. CDK5 has emerged as a crucial regulator of neuronal migration in the developing central nervous system. CDK5 phosphorylates a diverse list of substrates, implicating it in the regulation of a range of cellular processes - from adhesion and motility, to synaptic plasticity and drug addiction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDK5
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.07 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CDK5
Gene Accession No. P49615, Q00535, Q03114
Gene Alias AW048668; Cdk5; CDK5 activator 1; CDK5-SV; CDKN5; Cell division protein kinase 5; CR6 protein kinase; Crk6; cyclin dependent kinase 5; cyclin-dependent kinase 5; cyclin-dependent-like kinase 5; LIS7; protein kinase CDK5 splicing; PSSALRE; Serine/threonine-protein kinase PSSALRE; tau protein kinase II catalytic subunit; TPKII catalytic subunit
Gene Symbols CDK5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 170-289 of human CDK5 (NP_004926.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1020, 12568, 140908
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.