Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDK5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501726303 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-6303 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-6303 Supplier Proteintech Group Inc Supplier No. 104301AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cyclin-dependent kinase-5 (CDK5) is a member of the cyclin-dependent kinase family of serine/threonine kinases. Its mRNA and protein are expressed in the kidneys, testes, and ovaries. And Its activity has been detected almost exclusively in brain extracts. Similar to other Cdks, monomeric Cdk5 displays no enzymatic activity; however, Cdk5 is not activated bycyclins. Instead, Cdk5 activity requires association with one of two brain-specific regulatory subunits, called p35 and p39. These two activators regulate the spatial and temporal expression of active Cdk5, restricting its activity primarily to post-mitotic neurons.

Specifications

Antigen CDK5
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CDK5
Gene Accession No. P49615, Q00535, Q03114
Gene Alias CDK5, Cell division protein kinase 5, cyclin dependent kinase 5, PSSALRE, TPKII catalytic subunit
Gene Symbols Cdk5
Host Species Rabbit
Immunogen CDK5 Fusion Protein Ag0686
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1020, 12568, 140908
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.