Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDK5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980779
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0779 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0779 Supplier Bioss Supplier No. BS10258R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cyclin-dependent kinase-5 (CDK5) is a member of the cyclin-dependent kinase family of serine/threonine kinases. Its mRNA and protein are expressed in the kidneys, testes, and ovaries. And Its activity has been detected almost exclusively in brain extracts. Similar to other Cdks, monomeric Cdk5 displays no enzymatic activity; however, Cdk5 is not activated bycyclins. Instead, Cdk5 activity requires association with one of two brain-specific regulatory subunits, called p35 and p39. These two activators regulate the spatial and temporal expression of active Cdk5, restricting its activity primarily to post-mitotic neurons.

Specifications

Antigen CDK5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CDK5
Gene Accession No. Q00535
Gene Alias AW048668; CDK5; CDK5 activator 1; CDK5-SV; CDKN5; Cell division protein kinase 5; CR6 protein kinase; Crk6; cyclin dependent kinase 5; cyclin-dependent kinase 5; Cyclin-dependent-like kinase 5; LIS7; protein kinase CDK5 splicing; PSSALRE; Serine/threonine-protein kinase PSSALRE; Tau protein kinase II catalytic subunit; TPKII catalytic subunit
Gene Symbols CDK5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CDK5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1020
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.