Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDK5RAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590755
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590755 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590755 Supplier Invitrogen™ Supplier No. PA590755
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MHPLQCVLQV QRSLGWGPLA SVSWLSLRMC RAHSSLSSTM CPSPERQEDG ARKDFSSRLA AGPTFQHFLK SASAPQEKLS SEVEDPPPYL MMDELLGRQR KVYLETYGCQ MNVNDTEIAW SILQKSGYLR TSNLQEADVI LLVTCSIREK AEQTIWNRLH QLKALKTRRP RSRVPLRIGI LGCMAERLKE EILNREKMVD ILAGPDAYRD LPRLLAVAES GQQAANVLLS LDETYADVMP VQTSASATSA FVSIMRGCDN MCSYCIVPFT RGRERSRPIA SILEEVKKLS EQGLKEVTLL GQNVNSFRDN SEVQFNSAVP TNLSRGFTTN YKTKQGGLRF AHLLDQVSRV; Positive Samples: A-549, Mouse thymus.

Neuronal CDC2-like kinase, which is involved in the regulation of neuronal differentiation, is composed of a catalytic subunit, CDK5, and an activating subunit, p25NCK5A. The protein encoded by this gene binds to p25NCK5A and therefore may be involved in neuronal differentiation. Multiple transcript variants exist for this gene, but the full-length natures of only two have been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDK5RAP1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CDK5RAP1
Gene Accession No. Q8BTW8, Q96SZ6, Q9JLH6
Gene Alias 2310066P17Rik; C20orf34; C42; CDK5 activator-binding protein C42; CDK5 regulatory subunit associated protein 1; CDK5 regulatory subunit-associated protein 1; CDK5 regulatory subunit-associated protein 1-like protein; CDK5RAP1; CDK5RAP1.3; CDK5RAP1.4; CGI-05; HSPC167; mitochondrial tRNA methylthiotransferase CDK5RAP1; CDK5 regulatory subunit-associated protein 1
Gene Symbols CDK5RAP1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-350 of human CDK5RAP1 (NP_0574922).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 252827, 51654, 66971
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.