Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDK5RAP2 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100767AF647
Description
CDK5RAP2 Polyclonal antibody specifically detects CDK5RAP2 in Human samples. It is validated for Western BlotSpecifications
CDK5RAP2 | |
Polyclonal | |
Western Blot | |
C48, CDK5 activator-binding protein C48, CDK5 regulatory subunit associated protein 2, CDK5 regulatory subunit-associated protein 2, centrosomal protein 215 kDa, Centrosome-associated protein 215, Cep215, DKFZp686B1070, DKFZp686D1070, FLJ10867, KIAA1633microcephaly, primary autosomal recessive 3, MCPH3 | |
The immunogen recognized by this antibody maps to a region between residues 900 and 950 of human CDK5 Regulatory Subunit Associated Protein 2 using the numbering given in entry NP_060719.4 (GeneID 55755). | |
0.1 mL | |
Cell Cycle and Replication, Cellular Markers, Chromatin Research | |
55755 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction