Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDK5RAP2 Monoclonal Antibody (527CT22.9.2)
GREENER_CHOICE

Catalog No. PIMA537609
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537609 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537609 Supplier Invitrogen™ Supplier No. MA537609
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CDK5RAP2 Monoclonal Antibody for Western Blot

CDK5RAP2 is a regulator of CDK5 (cyclin-dependent kinase 5) activity. The protein is localized to the centrosome and Golgi complex and interacts with CDK5R1 and pericentrin (PCNT). CDK5RAP2 plays a role in centriole engagement and microtubule nucleation, and has been linked to primary microcephaly and Alzheimer's disease. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDK5RAP2
Applications Western Blot
Classification Monoclonal
Clone 527CT22.9.2
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.09% sodium azide; pH 7.4
Gene CDK5RAP2
Gene Accession No. Q96SN8
Gene Alias C48; CDK5 activator-binding protein C48; CDK5 regulatory subunit associated protein 2; CDK5 regulatory subunit-associated protein 2; CDK5RAP2; centrosomal protein 215 kDa; Centrosome-associated protein 215; centrosomin; Cep215; KIAA1633; MCPH3
Gene Symbols CDK5RAP2
Host Species Mouse
Immunogen recombinant protein of human CDK5RAP2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55755
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.