Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CDK5RAP2 Monoclonal Antibody (CL3392)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA535013
Description
Immunogen sequence: ELLEKLEKLF LNGKSVGVEM NTQNELMERI EEDNLTYQHL LPESPEPSAS HALSDYETSE KSFFSRDQKQ DNETEKTSVM V.
CDK5RAP2 is a regulator of CDK5 (cyclin-dependent kinase 5) activity. The protein is localized to the centrosome and Golgi complex and interacts with CDK5R1 and pericentrin (PCNT). CDK5RAP2 plays a role in centriole engagement and microtubule nucleation, and has been linked to primary microcephaly and Alzheimer's disease. Alternative splicing results in multiple transcript variants.
Specifications
CDK5RAP2 | |
Monoclonal | |
0.50 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96SN8 | |
CDK5RAP2 | |
Recombinant Protein corresonding to Human CDK5RAP2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
CL3392 | |
Unconjugated | |
CDK5RAP2 | |
C48; CDK5 activator-binding protein C48; CDK5 regulatory subunit associated protein 2; CDK5 regulatory subunit-associated protein 2; CDK5RAP2; centrosomal protein 215 kDa; Centrosome-associated protein 215; centrosomin; Cep215; KIAA1633; MCPH3 | |
Mouse | |
Protein A | |
RUO | |
55755 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction