Learn More
CDK5RAP2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300554AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Chimpanzee
CDK5RAP2 is a regulator of CDK5 (cyclin-dependent kinase 5) activity. The protein is localized to the centrosome and Golgi complex and interacts with CDK5R1 and pericentrin (PCNT). CDK5RAP2 plays a role in centriole engagement and microtubule nucleation, and has been linked to primary microcephaly and Alzheimer′s disease. Alternative splicing results in multiple transcript variants.Specifications
| CDK5RAP2 | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q96SN8 | |
| CDK5RAP2 | |
| Between 900 and 950 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| CDK5RAP2 | |
| CEP215, KIAA1633 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55755 | |
| 4° C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.