Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDK5RAP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545257
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545257 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545257 Supplier Invitrogen™ Supplier No. PA545257
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: IDWGDFGVEA VSEGTDSGIS AEAAGIDWGI FPESDSKDPG GDGIDWGDDA Sequence homology: Human: 100%.

This gene encodes a protein that has been reported to function in signaling pathways governing transcriptional regulation and cell cycle progression. It may play a role in tumorigenesis and metastasis. A pseudogene of this gene is located on the long arm of chromosome 20. Alternative splicing results in multiple transcript variants that encode different isoforms. [provided by RefSeq, May 2013].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDK5RAP3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CDK5RAP3
Gene Accession No. Q96JB5
Gene Alias 1810007E24Rik; BC002318; C53; C81486; CDK5 activator-binding protein C53; CDK5 activator-binding protein C53; CDK5 regulatory subunit associated protein 3; CDK5 regulatory subunit associated protein IC53-2; CDK5 regulatory subunit-associated protein 3; Cdk5rap3; HSF-27; IC53; IC53-2; ischemic heart CDK5 activator-binding protein C53; LXXLL/leucine-zipper-containing ARFbinding protein; LXXLL/leucine-zipper-containing ARF-binding protein; LZAP; MST016; MSTP016; OK/SW-cl.114; PP1553; Protein HSF-27
Gene Symbols CDK5RAP3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human CDK5RAP3 (aa 267-316).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80279
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.