Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDK5RAP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120895
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120895 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120895 Supplier Invitrogen™ Supplier No. PA5120895
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: U-87MG, 293T, LO2, A549, HT-29, Mouse brain, Mouse kidney, Mouse liver, Rat brain. The target is usually found in the following locations: Cytoplasm, Nucleus, centrosome, cytoskeleton, microtubule organizing center. Immunogen sequence: MEDHQHVPID IQTSKLLDWL VDRRHCSLKW QSLVLTIREK INAAIQDMPE SEEIAQLLSG SYIHYFHCLR ILDLLKGTEA STKNIFGRYS SQRMKDWQEI IALYEKDNTY LVELSSLLVR NVNYEIPSLK KQIAKCQQLQ QEYSRKEEEC QAGAAEMREQ FYHSCKQYGI TGENVRGELL ALVKDLPSQL AEIGAAAQQS LGEAIDVYQA SVGFVCESPT EQVLPMLRFV QKRGNSTVYE WRTGTEPSVV ERPHLEELPE QVAEDAIDWG DFGVEAVSEG TDSGISAEAA GIDWGIFPES.

This gene encodes a protein that has been reported to function in signaling pathways governing transcriptional regulation and cell cycle progression. It may play a role in tumorigenesis and metastasis. A pseudogene of this gene is located on the long arm of chromosome 20. Alternative splicing results in multiple transcript variants that encode different isoforms. [provided by RefSeq, May 2013].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDK5RAP3
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CDK5RAP3
Gene Accession No. Q96JB5, Q99LM2, Q9JLH7
Gene Alias 1810007E24Rik; BC002318; C53; C81486; CDK5 activator-binding protein C53; CDK5 activator-binding protein C53; CDK5 regulatory subunit associated protein 3; CDK5 regulatory subunit associated protein IC53-2; CDK5 regulatory subunit-associated protein 3; Cdk5rap3; HSF-27; IC53; IC53-2; ischemic heart CDK5 activator-binding protein C53; LXXLL/leucine-zipper-containing ARFbinding protein; LXXLL/leucine-zipper-containing ARF-binding protein; LZAP; MST016; MSTP016; OK/SW-cl.114; PP1553; Protein HSF-27
Gene Symbols CDK5RAP3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human CDK5RAP3 (NP_788276.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80278, 80279, 80280
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.