Learn More
Promega CDK6/CyclinD3 Kinase Enzyme System, 1mg
Catalog No. PRV6282 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6282 1 mg
1 options
Catalog No. PRV6282 Supplier Promega Supplier No. V6282
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human CDK6 and CyclinD3 were co-expressed by baculovirus in Sf9 insect cells using an N-terminal His tag on both proteins.
  • Easily Screen and Profile CDK6/CyclinD3 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for convenient bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C