Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CDK6 Recombinant Rabbit Monoclonal Antibody (0O6X7)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA535046
Description
Immunogen sequence: QLGKILDVIG LPGEEDWPRD VALPRQAFHS KSAQPIEKFV TDIDELGKDL LLKCLTFNPA KRISAYSALS HPYFQDLERC KENLDSHLPP SQNTSELNTA.
CDK6 functions as a cell-cycle initiator protein. This protein is coexpressed and copurified with CyclinD1. They are critical regulators of cell cycle progression. The prototype member of this family, p34cdc2, and a related protein cdk2, function late in the cycle while cdk4 and cdk6 are critically involved in G1 to S progression. CDK6 is essential for cell proliferation within the dentate gyrus of the hippocampus and the cuventricular zone of the lateral ventricles. Mutations in the gene results in microcephaly 12, primary, autosomal recessive (MCPH12).
Specifications
CDK6 | |
Recombinant Monoclonal | |
0.25 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q00534 | |
CDK6 | |
A synthetic peptide corresponding to a sequence within amino acids 227-326 of human CDK6 (Q00534). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
ELISA, Western Blot, Immunocytochemistry | |
0O6X7 | |
Unconjugated | |
CDK6 | |
5830411I20; AI504062; CDK6; CDKN6; Cell division protein kinase 6; CR2 protein kinase; Crk2; cyclin dependent kinase 6; cyclin-dependent kinase 6; MCPH12; MGC59692; PLSTIRE; Serine/threonine-protein kinase PLSTIRE | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1021, 114483 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction