Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cdk6 Rabbit anti-Human, Clone: S03-2G5, Novus Biologicals™

Rabbit Monoclonal Antibody
$263.00 - $573.00
Specifications
Antigen | Cdk6 |
---|---|
Clone | S03-2G5 |
Dilution | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence 20 μg/mL |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Description
Cdk6 Monoclonal antibody specifically detects Cdk6 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, ImmunofluorescenceSpecifications
Cdk6 | |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence 20 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Cell division protein kinase 6, cyclin-dependent kinase 6, EC 2.7.11, EC 2.7.11.22, PLSTIREMGC59692, Serine/threonine-protein kinase PLSTIRE | |
A synthetic peptide of human Cdk6 (Uniprot#: Q00534) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
S03-2G5 | |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication, Ovarian Carcinoma Cell Markers | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
1021 | |
IgG | |
Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title