Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cdk7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP32122525UL
Description
Cdk7 Polyclonal antibody specifically detects Cdk7 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
Cdk7 | |
Polyclonal | |
Western Blot 0.04-0.4 ug/ml, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
CAK1Xenopus laevis, cdk-activating kinase), CDK-activating kinase, Cell division protein kinase 7, cyclin-dependent kinase 7, cyclin-dependent kinase 7 (homolog of Xenopus MO15 cdk-activating kinase), EC 2.7.11, EC 2.7.11.22, EC 2.7.11.23, EC 2.7.11.24, p39 Mo15, serine/threonine kinase stk1,39 kDa protein kinase, serine/threonine protein kinase 1, serine/threonine protein kinase MO15, TFIIH basal transcription factor complex kinase subunit | |
This antibody was developed against Recombinant Protein corresponding to amino acids: GCILAELLLRVPFLPGDSDLDQLTRIFETLGTPTEEQWPDMCSLPDYVTFKSFPGIPLHHIFSAAGDDLLDLIQGLFLFNPCARITATQALKMKYFSNRPGPTPGCQLPRPNCPVETLKEQSNPALAIKRKRTEALEQGGLPKKLI | |
25 μg | |
Cell Cycle and Replication, DNA Repair, Nucleotide Excision Repair, Signal Transduction, Transcription Factors and Regulators | |
1022 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction