Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDK7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582321
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582321 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582321 Supplier Invitrogen™ Supplier No. PA582321
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GCILAELLLR VPFLPGDSDL DQLTRIFETL GTPTEEQWPD MCSLPDYVTF KSFPGIPLHH IFSAAGDDLL DLIQGLFLFN PCARITATQA LKMKYFSNRP GPTPGCQLPR PNCPVETLKE QSNPALAIKR KRTEALEQGG LPKKLI Highest antigen sequence indentity to the following orthologs: Rat - 92%, Mouse - 89%.

The trimeric complex of CDK7, cyclin H and MNAT1, functions as a Cdk-activating kinase (CAK). It is an essential component of the transcription factor TFIIH, that is involved in transcription initiation and DNA repair. This complex is thought to serve as a direct link between the regulation of transcription and the cell cycle. The bound complex is important in the normal double-strand break repair function of breast cancer, type 1 (BRCA1). CDK7 may be a target of germline cancer-inducing mutations.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDK7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDK7
Gene Accession No. P50613
Gene Alias 39 KDa protein kinase; 39 protein kinase; AI323415; AI528512; C230069N13; CAK; CAK1; CDK; CDK7; CDK-activating kinase; CDK-activating kinase 1; Cdkn7; cell division protein kinase 7; CR4 protein kinase; CRK4; CRK4 PK (CDC2-related-kinase-4 protein kinase); cyclin dependent kinase 7; cyclin-dependent kinase 7; cyclin-dependent kinase 7 (homolog of Xenopus MO15 cdk-activating kinase); cyclin-dependent kinase 7 (MO15 homolog Xenopus laevis cdk-activating kinase); cyclin-dependent kinase 7 (MO15 homolog, Xenopus laevis, cdk-activating kinase); EC 2.7.11.22; EC 2.7.11.23; HCAK; homolog of Xenopus MO15 Cdk-activating kinase; kinase subunit of CAK; MO15; Mpk-7; OTTHUMP00000222032; OTTHUMP00000222033; p39 Mo15; p39MO15; Protein-tyrosine kinase MPK-7; serine/threonine kinase stk1; serine/threonine protein kinase 1; serine/threonine protein kinase MO15; Serine/threonine-protein kinase 1; STK1; TFIIH basal transcription factor complex kinase subunit
Gene Symbols CDK7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CDK7. Recombinant protein control fragment (Product #RP-102518).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1022
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.