Learn More
CDK7 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$167.95
Specifications
| Antigen | CDK7 |
|---|---|
| Applications | Immunocytochemistry, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-155-7547
|
Bethyl Laboratories, Inc
A300405AT |
20 μL |
Each of 1 for $167.95
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
The protein encoded by this gene is a member of the cyclin-dependent protein kinase (CDK) family. CDK family members are highly similar to the gene products of Saccharomyces cerevisiae cdc28, and Schizosaccharomyces pombe cdc2, and are known to be important regulators of cell cycle progression. This protein forms a trimeric complex with cyclin H and MAT1, which functions as a Cdk-activating kinase (CAK). It is an essential component of the transcription factor TFIIH, that is involved in transcription initiation and DNA repair. This protein is thought to serve as a direct link between the regulation of transcription and the cell cycle.Specifications
| CDK7 | |
| Polyclonal | |
| Liquid | |
| RUO | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| EC 2.7.11.22, EC 2.7.11.23, CDK-activating kinase, CAK, TFIIH basal transcription factor complex kinase subunit | |
| Cdk7 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
| Immunocytochemistry, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P50613, P51952, Q03147 | |
| 1022, 12572, 171150 | |
| Between 300 and C-term | |
| Primary | |
| 4° C | |
| CDK7 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.