Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CDK8/19i
GREENER_CHOICESDP

Catalog No. 737250 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
73-725-0 50 mg
73-721-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 73-725-0 Supplier Tocris Bioscience™ Supplier No. 7372/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective CDK8 and CDK19 inhibitor; maintains pluripotency of mouse PSCs in culture

CDK8/19i is a potent and selective CDK8 and CDK19 inhibitor (IC50 values are 2.9 and 14.1 nM, respectively). CDK8/19i maintains pluripotency of mouse PSCs in culture. It induces the formation of dome-shaped colonies and upregulates Nanog-GFP reporter and endogenous Nanog expression in mouse ESCs in vitro.

Specifications

Chemical Name or Material N-Propyl-3-(4-pyridinyl)imidazo[1,2-b]pyridazin-6-amine
CAS 923596-52-5
Quantity 50 mg
Target Cyclin-dependent Kinase Inhibitors
Molecular Formula C14H15N5
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.