Learn More
Promega CDK9/CyclinK Kinase Enzyme System, 10microg
Catalog No. PRV4104 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4104 10 μg
1 options
Catalog No. PRV4104 Supplier Promega Supplier No. V4104
Recombinant full-length human CDK9 and CyclinK proteins were co-expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CDK9/CyclinK is a member of the cyclin-dependent protein kinase (CDK) family.
  • Easily Screen and Profile CDK9/CyclinK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C