Learn More
Promega CDK9/CyclinK Kinase Enzyme System, 1mg
Catalog No. PRV6283 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6283 1 mg
1 options
Catalog No. PRV6283 Supplier Promega Supplier No. V6283
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human CDK9 and CyclinK proteins were co-expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CDK9/CyclinK is a member of the cyclin-dependent protein kinase (CDK) family.
  • Easily Screen and Profile CDK9/CyclinK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for convenient bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C