Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cdk9, p Thr29 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177756
Description
Cdk9 Polyclonal specifically detects Cdk9 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
Cdk9 (p Thr29) | |
Polyclonal | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
C-2k, CDC2L4CTK1, CDC2-related kinase, Cell division cycle 2-like protein kinase 4, Cell division protein kinase 9, cyclin-dependent kinase 9, cyclin-dependent kinase 9 (CDC2-related kinase), EC 2.7.11, EC 2.7.11.22, EC 2.7.11.23, PITALRE, serine/threonine protein kinase PITALRE, Serine/threonine-protein kinase PITALRE, TAK | |
Rabbit | |
42.78 kDa | |
0.1 mg | |
Cancer, Cell Cycle and Replication, Phospho Specific, Signal Transduction, Virology Bacteria and Parasites | |
1025 | |
Human, Mouse, Rat | |
IgG |
Western Blot, ELISA, Immunoprecipitation | |
1.3 mg/ml | |
Western Blot 1:200-1:2000, ELISA 1:5000-1:24000, Immunoprecipitation 1:100 | |
NC_000009.11 | |
CDK9 | |
This affinity purified Cdk9 [p Thr29] Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to residues surrounding T29 in the human Cdk9 protein. (Uniprot: P50750) | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is specific for human CDK9 protein phosphorylated at T29. A BLAST analysis was used to suggest cross-reactivity with CDK9 from human, mouse and rat based on 100% homology with the immunizing sequence. Cross-reactivity with CDK9 from other sources has not been determined. | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction