Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdk9 Polyclonal Antibody, Abbomax

Catalog No. 502204068
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4068 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4068 Supplier Abbomax Inc Supplier No. 602390
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Human pancreas, pancreatic cancer Cellular location: Nucleus.

Cdk9 Polyclonal antibody specifically detects Cdk9 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Cdk9
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene CDK9
Gene Accession No. P50750, Q99J95
Gene Alias C-2k; CDC2L4; CDC2-related kinase; Cdk9; Cell division cycle 2-like protein kinase 4; Cell division protein kinase 9; CTK1; cyclin dependent kinase 9; cyclin-dependent kinase 9; cyclin-dependent kinase 9 (CDC2-related kinase); PITALRE; RP11-228B15.5; serine/threonine protein kinase PITALRE; Serine/threonine-protein kinase PITALRE; TAK; Tat-associated kinase complex catalytic subunit
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the C-terminus of mouse PLAC1 protein (from 145aa-175aa)
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1025, 107951
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.