Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDK9 Polyclonal Antibody, Active Motif™

Rabbit Polyclonal Antibody
$156.65 - $620.58
Specifications
Antigen | CDK9 |
---|---|
Concentration | Lot-Specific |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3497
![]() ![]() |
Active Motif
61560 |
10 μL |
Each for $156.65
|
|
|||||
50-199-3496
![]() ![]() |
Active Motif
61559 |
100 μL |
Each for $620.58
|
|
|||||
Description
CDK9 (cyclin-dependent kinase 9) is a member of the cyclin-dependent protein kinase family. It is involved in the regulation of transcription. CDK9 is a member of the cyclin-dependent kinase pair (CDK9/cyclin-T) complex. This complex facilitates the transition from abortive to productive elongation by phosphorylating the CTD of the large subunit of RNA polymerase II, POLR2A, SUPT5H, and RDBP. The complex is inactive when in the 7SK snRNP complex form. CDK9 phosphorylates EP300, MYOD1, RPB1/POLR2A, AR, DSIF, and NELF. It regulates cytokine inducible transcription networks by facilitating promoter recognition of target transcription factors. CDK9 promotes RNA synthesis in genetic programs for cell growth, differentiation, and viral pathogenesis. The CDK9/cyclin-K complex also has kinase activity towards CTD of RNAPII and can substitute for CDK9/cyclin-T P-TEFb in vitro. Chronic activation of CDK9 causes cardiac myocyte enlargement leading to cardiac hypertrophy and confers a predisposition to heart failure.Specifications
CDK9 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P50750 | |
1025 | |
This antibody was raised against a peptide within the N-terminal region of human CDk9. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
CDK9 |
Lot-Specific | |
Polyclonal | |
Liquid | |
RUO | |
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8 | |
BOS_12097; C-2K; CDC2L4; CDC2-related kinase; CDK9; Cell division cycle 2-like protein kinase 4; Cell division protein kinase 9; CTK1; cyclin dependent kinase 9; Cyclin-dependent kinase 9; cyclin-dependent kinase 9 (CDC2-related kinase); PITALRE; RP11-228B15.5; serine/threonine protein kinase PITALRE; Serine/threonine-protein kinase PITALRE; TAK; Tat-associated kinase complex catalytic subunit | |
CDK9 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title