Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDK9 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983156
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3156 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3156 Supplier Bioss Supplier No. BS1388R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CDK9 (cyclin-dependent kinase 9) is a member of the cyclin-dependent protein kinase family. It is involved in the regulation of transcription. CDK9 is a member of the cyclin-dependent kinase pair (CDK9/cyclin-T) complex. This complex facilitates the transition from abortive to productive elongation by phosphorylating the CTD of the large subunit of RNA polymerase II, POLR2A, SUPT5H, and RDBP. The complex is inactive when in the 7SK snRNP complex form. CDK9 phosphorylates EP300, MYOD1, RPB1/POLR2A, AR, DSIF, and NELF. It regulates cytokine inducible transcription networks by facilitating promoter recognition of target transcription factors. CDK9 promotes RNA synthesis in genetic programs for cell growth, differentiation, and viral pathogenesis. The CDK9/cyclin-K complex also has kinase activity towards CTD of RNAPII and can substitute for CDK9/cyclin-T P-TEFb in vitro. Chronic activation of CDK9 causes cardiac myocyte enlargement leading to cardiac hypertrophy and confers a predisposition to heart failure.

Specifications

Antigen CDK9
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CDK9
Gene Accession No. P50750
Gene Alias BOS_12097; C-2K; CDC2L4; CDC2-related kinase; CDK9; Cell division cycle 2-like protein kinase 4; Cell division protein kinase 9; CTK1; cyclin dependent kinase 9; Cyclin-dependent kinase 9; cyclin-dependent kinase 9 (CDC2-related kinase); PITALRE; RP11-228B15.5; serine/threonine protein kinase PITALRE; Serine/threonine-protein kinase PITALRE; TAK; Tat-associated kinase complex catalytic subunit
Gene Symbols CDK9
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CDK9.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1025
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.