Learn More
Invitrogen™ CDKAL1 Polyclonal Antibody

Description
The antibody detects endogenous levels of total CDKAL1 protein.
Specifications
Specifications
| Antigen | CDKAL1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Cdkal1 |
| Gene Accession No. | Q5VV42 |
| Gene Alias | 1190005B03Rik; 6620401C13Rik; CDK5 regulatory subunit associated protein 1 like 1; CDK5 regulatory subunit associated protein 1-like 1; CDK5 regulatory subunit-associated protein 1-like 1; CDKAL; CDKAL1; Threonylcarbamoyladenosine tRNA methylthiotransferase; tRNA-t(6)A37 methylthiotransferase |
| Gene Symbols | Cdkal1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.