Learn More
Invitrogen™ CDKAL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109506
Description
Immunogen sequence: MPSASCDTLL DDIEDIVSQE DSKPQDRHFV RKDVVPKVRR RNTQKYLQEE ENSPPSDSTI PGIQKIWIRT WGCSHNNSDG EYMAGQLAAY GYKITENASD ADLWLLNSCT VKNPAEDHFR NSIKKAQEEN KKIVLAGCVP QAQPRQDYLK GLSIIGVQQI DRVVEVVEET IKGHSVRLLG QKKDNGRRLG GARLDLPKIR KNPLIEIISI NTGCLNACTY CKTKHARGNL.
The protein encoded by this gene is a member of the methylthiotransferase family. The function of this gene is not known. Genome-wide association studies have linked single nucleotide polymorphisms in an intron of this gene with susceptibility to type 2 diabetes.
Specifications
CDKAL1 | |
Polyclonal | |
Unconjugated | |
Cdkal1 | |
1190005B03Rik; 6620401C13Rik; CDK5 regulatory subunit associated protein 1 like 1; CDK5 regulatory subunit associated protein 1-like 1; CDK5 regulatory subunit-associated protein 1-like 1; CDKAL; CDKAL1; Threonylcarbamoyladenosine tRNA methylthiotransferase; tRNA-t(6)A37 methylthiotransferase | |
Rabbit | |
Affinity Chromatography | |
RUO | |
54901, 68916 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
0.64 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5VV42, Q91WE6 | |
Cdkal1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-230 of human CDKAL1 (NP_060244.2). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.